Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-5-chloro-2-methoxybenzoic acid, 98+%

Catalog No. AAB2428106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2428106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2428106 Supplier Thermo Scientific Chemicals Supplier No. B2428106

CAS: 7206-70-4 | C8H8ClNO3 | 201.606 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7206-70-4
Molecular Formula C8H8ClNO3
Molecular Weight (g/mol) 201.606
MDL Number MFCD00038794
InChI Key RVEATKYEARPWRE-UHFFFAOYSA-N
Synonym 4-amino-5-chloro-o-anisic acid, unii-pq8l2l84el, benzoic acid, 4-amino-5-chloro-2-methoxy, pq8l2l84el, pubchem15052, acmc-209oll, metoclopramide impurity c, 2-methoxy-4-amino-5-chlorobenzoic acid, 4amino-5-chloro-2-methoxy-benzoic acid, 5-chloro-4-amino-2-methoxybenzoic acid
PubChem CID 81626
IUPAC Name 4-amino-5-chloro-2-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1C(=O)O)Cl)N

Specifications

Melting Point 205°C to 208°C (decomposition)
Quantity 5 g
Formula Weight 201.61
Percent Purity ≥98%
Chemical Name or Material 4-Amino-5-chloro-2-methoxybenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 230-582-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.