Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-5-chloro-o-anisic Acid 98.0+%, TCI America™
SDP

Catalog No. A1237500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1237500G 500 g
A123725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1237500G Supplier TCI America Supplier No. A1237500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7206-70-4
Molecular Formula C8H8ClNO3
Molecular Weight (g/mol) 201.606
MDL Number MFCD00038794
InChI Key RVEATKYEARPWRE-UHFFFAOYSA-N
Synonym 4-amino-5-chloro-o-anisic acid, unii-pq8l2l84el, benzoic acid, 4-amino-5-chloro-2-methoxy, pq8l2l84el, pubchem15052, acmc-209oll, metoclopramide impurity c, 2-methoxy-4-amino-5-chlorobenzoic acid, 4amino-5-chloro-2-methoxy-benzoic acid, 5-chloro-4-amino-2-methoxybenzoic acid
PubChem CID 81626
IUPAC Name 4-amino-5-chloro-2-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1C(=O)O)Cl)N

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 201.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-5-chloro-o-anisic Acid
EINECSNumber (9)-0076
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.