Learn More
Selleck Chemical LLC 4-Amino-5-imidazolecarboxamide S3651-25mg
SDP
Catalog No. 503109150
Encompass_Preferred
4-Amino-5-imidazolecarboxamide is an imidazole derivative which is a metabolite of the antineoplastic agents BIC and DIC By itself or as the ribonucleotide it is used as a condensation agent in the preparation of nucleosides and nucleotides
Catalog No. 50-310-9150 Supplier Selleck Chemical LLC Supplier No. S365125MG
4-Amino-5-imidazolecarboxamide is an imidazole derivative which is a metabolite of the antineoplastic agents BIC and DIC By itself or as the ribonucleotide it is used as a condensation agent in the preparation of nucleosides and nucleotides