Learn More
Medchemexpress LLC 4-amino-6-chloro-1,3-benzenedisulfonamide | 121-30-2 | MFCD00007933 | 99.6% | 285.73 g/mol | C6H8ClN3O4S2 | 100 G
SDP
Catalog No. 5000301577
Encompass_Preferred
Chloraminophenamide (4-amino-6-chloro-1,3-benzenedisulfonamide; CAS 121-30-2) is a carbonic anhydrase inhibitor provided for preclinical research, including antiglaucoma studies and enzymology. It is supplied as a high-purity solid intended for biochemical assays, structure-activity studies, and other laboratory applications.
  • High purity (≈99.6%) suitable for research assays.
  • Well-characterized molecular weight of 285.73 g/mol.
  • Chemical formula C6H8ClN3O4S2 for accurate stoichiometry.
  • Stable when stored at 4°C; protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Useful as a tool compound for carbonic anhydrase inhibition and antiglaucoma research.
Catalog No. 50-003-01577 Supplier Medchemexpress LLC Supplier No. HYW031519100G
Chloraminophenamide (4-amino-6-chloro-1,3-benzenedisulfonamide; CAS 121-30-2) is a carbonic anhydrase inhibitor provided for preclinical research, including antiglaucoma studies and enzymology. It is supplied as a high-purity solid intended for biochemical assays, structure-activity studies, and other laboratory applications.
  • High purity (≈99.6%) suitable for research assays.
  • Well-characterized molecular weight of 285.73 g/mol.
  • Chemical formula C6H8ClN3O4S2 for accurate stoichiometry.
  • Stable when stored at 4°C; protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Useful as a tool compound for carbonic anhydrase inhibition and antiglaucoma research.