Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-6-hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 90+%, Thermo Scientific™

Catalog No. AAL0369106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0369106 5 g
AAL0369103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0369106 Supplier Thermo Scientific Chemicals Supplier No. L0369106

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5472-41-3
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.129
MDL Number MFCD00005689
InChI Key KTQYLKORCCNJTQ-UHFFFAOYSA-N
Synonym 4-amino-6-hydroxypyrazolo 3,4-d pyrimidine, 4-amino-1h-pyrazolo 3,4-d pyrimidin-6-ol, 4-aminopyrazolo 3,4-d pyrimidin-6-ol, 4-amino-2h-pyrazolo 3,4-d pyrimidin-6-ol, 4-amino-1h-pyrazolo 3,4-d pyrimidin-6 7h-one, 4-aminopyrazolo 5,4-d pyrimidin-6-ol, acmc-1akdw, 4-amino-6-hydroxypyrazolo-3,4-d pyrimidine, 4-amino-1h-pyrazolo 4,3-e pyrimidin-6-ol, 4-amino-7-hydroxypyrazolo 3,4-d pyrimidine
PubChem CID 79623
IUPAC Name 4-amino-1,2-dihydropyrazolo[3,4-d]pyrimidin-6-one
SMILES C1=C2C(=NC(=O)N=C2NN1)N

Specifications

Melting Point >300°C
Odor Odorless
Quantity 5 g
Beilstein 152192
Formula Weight 151.13
Percent Purity ≥90%
Chemical Name or Material 4-Amino-6-hydroxy-1H-pyrazolo[3,4-d]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.