Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-exatecan-d5 | 00-00-0 | 98.6% | C23H16D5N3O4 | 10MG
SDP

Catalog No. 5000260356
Encompass_Preferred

(4-NH2)-Exatecan-d5 is the deuterium-labeled form of (4-amino) exatecan, a topoisomerase inhibitor derivative of exatecan intended for research use. It is commonly used as a building block in antibody-drug conjugate synthesis and as a labeled tracer or internal standard for quantitative analytical methods.

  • Deuterium-labeled analog for analytical studies.
  • Suitable for synthesis of antibody-drug conjugates.
  • Usable as tracer or internal standard for NMR, GC-MS, and LC-MS.
  • High purity (98.6%) for reproducible results.
  • Molecular formula C23H16D5N3O4; molecular weight 408.46.

Catalog No. 50-002-60356 Supplier Medchemexpress LLC Supplier No. HY145397S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(4-NH2)-Exatecan-d5 is the deuterium-labeled form of (4-amino) exatecan, a topoisomerase inhibitor derivative of exatecan intended for research use. It is commonly used as a building block in antibody-drug conjugate synthesis and as a labeled tracer or internal standard for quantitative analytical methods.

  • Deuterium-labeled analog for analytical studies.
  • Suitable for synthesis of antibody-drug conjugates.
  • Usable as tracer or internal standard for NMR, GC-MS, and LC-MS.
  • High purity (98.6%) for reproducible results.
  • Molecular formula C23H16D5N3O4; molecular weight 408.46.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.