Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-L-phenylalanine | 943-80-6 | MFCD00069927 | 100.0% | 180.21 g/mol | C9H12N2O2 | 1 ML
SDP

Catalog No. 5000299361
Encompass_Preferred

4-Amino-L-phenylalanine (CAS 943-80-6) is an amino acid derivative and metabolic intermediate used in organic synthesis and in the production of some antibiotics. It is supplied for research use in both solid form and as a 10 mM aqueous stock solution. The compound has chemical formula C9H12N2O2 and a molecular weight of 180.21 g/mol.

  • Metabolic intermediate for antibiotic and organic synthesis.
  • High purity (99.98%).
  • Available as a 10 mM aqueous stock solution and solids (500 mg-5 g).
  • Chemical formula C9H12N2O2; molecular weight 180.21 g/mol.
  • Intended for research use as a reagent and building block.

Catalog No. 50-002-99361 Supplier Medchemexpress LLC Supplier No. HYW01648010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Amino-L-phenylalanine (CAS 943-80-6) is an amino acid derivative and metabolic intermediate used in organic synthesis and in the production of some antibiotics. It is supplied for research use in both solid form and as a 10 mM aqueous stock solution. The compound has chemical formula C9H12N2O2 and a molecular weight of 180.21 g/mol.

  • Metabolic intermediate for antibiotic and organic synthesis.
  • High purity (99.98%).
  • Available as a 10 mM aqueous stock solution and solids (500 mg-5 g).
  • Chemical formula C9H12N2O2; molecular weight 180.21 g/mol.
  • Intended for research use as a reagent and building block.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.