Learn More
Medchemexpress LLC 4-amino-N-(1-benzylpiperidin-4-yl)-5-chloro-2-methoxybenzamide;2-hydroxybutanedioic acid | 57645-91-7 | MFCD01743960 | 99.4% | 507.96 | C24H30ClN3O7 | 10 MG
SDP
Catalog No. 5000298027
Encompass_Preferred
Clebopride malate is the malate salt of clebopride, an orally active dopamine D2 receptor antagonist supplied as a research-grade analytical standard for laboratory pharmacology and analytical studies. It is provided as a high-purity solid intended for controlled experimental use and should be stored sealed and away from moisture; solutions may be frozen for extended storage.
  • Malate salt form of clebopride.
  • Orally active dopamine D2 receptor antagonist.
  • High purity suitable for analytical applications.
  • Supplied in a small research-grade pack for laboratory use.
  • Stable when stored sealed and away from moisture; follow refrigerated or frozen conditions for solutions.
Catalog No. 50-002-98027 Supplier Medchemexpress LLC Supplier No. HYB1613A10MG
Clebopride malate is the malate salt of clebopride, an orally active dopamine D2 receptor antagonist supplied as a research-grade analytical standard for laboratory pharmacology and analytical studies. It is provided as a high-purity solid intended for controlled experimental use and should be stored sealed and away from moisture; solutions may be frozen for extended storage.
  • Malate salt form of clebopride.
  • Orally active dopamine D2 receptor antagonist.
  • High purity suitable for analytical applications.
  • Supplied in a small research-grade pack for laboratory use.
  • Stable when stored sealed and away from moisture; follow refrigerated or frozen conditions for solutions.