Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-N-[3-methyl-1-[(6-methyl-2-pyridinyl)methyl]indazol-4-yl]thieno[3,2-d]pyrimidine-7-carboxamide | 1527517-50-5 | 98.6% | 429.5 g·mol-1 | C22H19N7OS | 100MG
SDP

Catalog No. 5000236211
Encompass_Preferred

c-Fms-IN-10 is a thienopyrimidine-derived small-molecule inhibitor of the c-Fms (colony stimulating factor-1 receptor, CSF-1R) receptor tyrosine kinase. It exhibits potent biochemical activity (reported IC50 ≈ 2 nM) and has demonstrated antitumor activity in preclinical studies. The compound is supplied as a solid for research use and is typically provided in milligram-scale quantities.

  • Potent c-Fms inhibition (IC50 ≈ 2 nM).
  • Molecular formula C22H19N7OS and molecular weight 429.5 g·mol-1.
  • High purity typically around 98.6%.
  • Soluble in DMSO for preparation of stock solutions.
  • Store powder at -20 °C to maintain stability.

Catalog No. 50-002-36211 Supplier Medchemexpress LLC Supplier No. HY126297100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

c-Fms-IN-10 is a thienopyrimidine-derived small-molecule inhibitor of the c-Fms (colony stimulating factor-1 receptor, CSF-1R) receptor tyrosine kinase. It exhibits potent biochemical activity (reported IC50 ≈ 2 nM) and has demonstrated antitumor activity in preclinical studies. The compound is supplied as a solid for research use and is typically provided in milligram-scale quantities.

  • Potent c-Fms inhibition (IC50 ≈ 2 nM).
  • Molecular formula C22H19N7OS and molecular weight 429.5 g·mol-1.
  • High purity typically around 98.6%.
  • Soluble in DMSO for preparation of stock solutions.
  • Store powder at -20 °C to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.