Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-N-[3-methyl-1-[(6-methyl-2-pyridinyl)methyl]indazol-4-yl]thienopyrimidine-7-carboxamide | 1527517-50-5 | 98.6% | 429.5 g/mol | C22H19N7OS | 25MG
SDP

Catalog No. 5000237388
Encompass_Preferred

c-Fms-IN-10 is a thieno[3,2-d]pyrimidine-derived small-molecule inhibitor of the receptor tyrosine kinase c-Fms (colony-stimulating factor-1 receptor, CSF-1R). It exhibits potent inhibitory activity against CSF-1R (reported IC50 = 2 nM), has demonstrated anti-tumor activity in preclinical studies, and is supplied as a high-purity solid or as a 10 mM solution in DMSO for biochemical and cellular research applications.

  • Molecular formula: C22H19N7OS.
  • Molecular weight: 429.5 g/mol.
  • Purity: 98.6% (as listed).
  • Reported potency: CSF-1R IC50 = 2 nM.
  • Available as powder and as a 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-37388 Supplier Medchemexpress LLC Supplier No. HY12629725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

c-Fms-IN-10 is a thieno[3,2-d]pyrimidine-derived small-molecule inhibitor of the receptor tyrosine kinase c-Fms (colony-stimulating factor-1 receptor, CSF-1R). It exhibits potent inhibitory activity against CSF-1R (reported IC50 = 2 nM), has demonstrated anti-tumor activity in preclinical studies, and is supplied as a high-purity solid or as a 10 mM solution in DMSO for biochemical and cellular research applications.

  • Molecular formula: C22H19N7OS.
  • Molecular weight: 429.5 g/mol.
  • Purity: 98.6% (as listed).
  • Reported potency: CSF-1R IC50 = 2 nM.
  • Available as powder and as a 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.