Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-N-(5-chloro-2-quinoxalinyl)-benzenesulfonamide | 97919-22-7 | 99.1% | 334.78 | C14H11ClN4O2S | 1 ML
SDP

Catalog No. 5000241095
Encompass_Preferred

Chloroquinoxaline sulfonamide is a small-molecule bioactive compound (CAS 97919-22-7) supplied for research use. It is available as a ready-to-use DMSO solution or as solid material for biochemical and cell-based assays.

  • High purity (99.1%).
  • Molecular weight 334.78 and formula C14H11ClN4O2S.
  • Offered as 10 mM, 1 mL solution in DMSO for immediate use.
  • Available as solid in multiple sizes (5 mg-100 mg) for flexible dosing.
  • Suitable for topoisomerase and parasitic research applications.
  • Supplied with batch certificate of analysis.

Catalog No. 50-002-41095 Supplier Medchemexpress LLC Supplier No. HY10666210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chloroquinoxaline sulfonamide is a small-molecule bioactive compound (CAS 97919-22-7) supplied for research use. It is available as a ready-to-use DMSO solution or as solid material for biochemical and cell-based assays.

  • High purity (99.1%).
  • Molecular weight 334.78 and formula C14H11ClN4O2S.
  • Offered as 10 mM, 1 mL solution in DMSO for immediate use.
  • Available as solid in multiple sizes (5 mg-100 mg) for flexible dosing.
  • Suitable for topoisomerase and parasitic research applications.
  • Supplied with batch certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.