Learn More
Medchemexpress LLC 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide | 144-82-1 | MFCD00053363 | 99.8% | 270.33 | C9H10N4O2S2 | 10 MG
SDP
Catalog No. 5000297079
Encompass_Preferred
Sulfamethizole (Standard) is an analytical reference standard of the sulfonamide antibiotic sulfamethizole, supplied for research and analytical applications. It is intended for use in qualitative and quantitative assays, method development, and instrument calibration with techniques such as HPLC, GC, and MS. For research use only; not for human or veterinary use.
  • High purity: 99.79%
  • Useful as a reference standard for HPLC, GC, and MS
  • Suitable for qualitative and quantitative assays
  • Compact 10 mg package for analytical workflows
  • Solid, white to off-white appearance for easy handling
Catalog No. 50-002-97079 Supplier Medchemexpress LLC Supplier No. HYB0333R10MG
Sulfamethizole (Standard) is an analytical reference standard of the sulfonamide antibiotic sulfamethizole, supplied for research and analytical applications. It is intended for use in qualitative and quantitative assays, method development, and instrument calibration with techniques such as HPLC, GC, and MS. For research use only; not for human or veterinary use.
  • High purity: 99.79%
  • Useful as a reference standard for HPLC, GC, and MS
  • Suitable for qualitative and quantitative assays
  • Compact 10 mg package for analytical workflows
  • Solid, white to off-white appearance for easy handling