Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-N-(tert-butoxycarbonyl)-L-phenylalanine 96.0+%, TCI America™
SDP

Catalog No. A21315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21315G 5 g
A21311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21315G Supplier TCI America Supplier No. A21315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55533-24-9
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00038139
InChI Key NDMVQEZKACRLDP-NSHDSACASA-N
Synonym boc-phe 4-nh2-oh, boc-4-amino-l-phenylalanine, boc-l-4-aminophenylalanine, 4-amino-n-boc-l-phenylalanine, s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 4-amino-n-tert-butoxycarbonyl-l-phenylalanine, 2s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-4-aminophenyl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-4-aminophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid
PubChem CID 2761482
IUPAC Name (2S)-3-(4-aminophenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(N)C=C1)C(O)=O

Specifications

Melting Point 126°C
Quantity 5 g
Formula Weight 280.32
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-N-(tert-butoxycarbonyl)-L-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.