Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-o-cresol 98.0+%, TCI America™
SDP

Catalog No. A15365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15365G 5 g
A153625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15365G Supplier TCI America Supplier No. A15365G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2835-96-3
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007878
InChI Key HDGMAACKJSBLMW-UHFFFAOYSA-N
Synonym 4-Amino-2-methylphenol, 4-Hydroxy-3-methylaniline
PubChem CID 76081
IUPAC Name 4-amino-2-methylphenol
SMILES CC1=C(C=CC(=C1)N)O

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-o-cresol
EINECSNumber (3)-0707
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.