Learn More
Medchemexpress LLC 4-Amino-TEMPO | 14691-88-4 | 171.26 | 1 ML
SDP
Catalog No. 5000445916
Encompass_Preferred
4-Amino-TEMPO, also known as 4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl, is a stable nitroxide radical and N-nucleophile derived from TEMPO. It exhibits superoxide dismutase-mimetic activity, offering protection to cells against oxidative damage and possessing radioprotective effects. This compound is extensively utilized across various fields, including biomedicine, materials science, and industrial production.
  • Serves as a spin label for detecting free radicals
  • Functions as an oxidation catalyst in industrial processes
  • Acts as an antioxidant stabilizer for polymers
  • Protects cells from oxidative damage
  • Exhibits radioprotective effects
  • Aids in studying the mechanism of oxidative stress
  • Can be detected with high sensitivity and selectivity using dihydroxycoumarins
Catalog No. 50-004-45916 Supplier Medchemexpress LLC Supplier No. HYW00200410MM/1ML
Add to Cart
Add to Cart
4-Amino-TEMPO, also known as 4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl, is a stable nitroxide radical and N-nucleophile derived from TEMPO. It exhibits superoxide dismutase-mimetic activity, offering protection to cells against oxidative damage and possessing radioprotective effects. This compound is extensively utilized across various fields, including biomedicine, materials science, and industrial production.
  • Serves as a spin label for detecting free radicals
  • Functions as an oxidation catalyst in industrial processes
  • Acts as an antioxidant stabilizer for polymers
  • Protects cells from oxidative damage
  • Exhibits radioprotective effects
  • Aids in studying the mechanism of oxidative stress
  • Can be detected with high sensitivity and selectivity using dihydroxycoumarins