Learn More
Medchemexpress LLC 4-Amino-TEMPO (4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl) | 14691-88-4 | 171.26 | 10 G
SDP
Catalog No. 5000447819
Encompass_Preferred
4-Amino-TEMPO is a stable nitroxide radical and N-nucleophile based on TEMPO. It exhibits superoxide dismutase-mimetic activity, protecting cells from oxidative damage, and offers radioprotective effects. It is widely used in biomedicine, materials science, and industrial production.
  • Stable nitroxide radical and N-nucleophile
  • Exhibits superoxide dismutase-mimetic activity
  • Protects cells from oxidative damage
  • Provides radioprotective effects
  • Used as a spin label for detecting free radicals
  • Functions as an oxidation catalyst
  • Acts as an antioxidant stabilizer for polymers
Catalog No. 50-004-47819 Supplier Medchemexpress LLC Supplier No. HYW00200410G
Add to Cart
Add to Cart
4-Amino-TEMPO is a stable nitroxide radical and N-nucleophile based on TEMPO. It exhibits superoxide dismutase-mimetic activity, protecting cells from oxidative damage, and offers radioprotective effects. It is widely used in biomedicine, materials science, and industrial production.
  • Stable nitroxide radical and N-nucleophile
  • Exhibits superoxide dismutase-mimetic activity
  • Protects cells from oxidative damage
  • Provides radioprotective effects
  • Used as a spin label for detecting free radicals
  • Functions as an oxidation catalyst
  • Acts as an antioxidant stabilizer for polymers