Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Aminoacetophenone Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111016027 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6027 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6027 Supplier MilliporeSigma Supelco Supplier No. PHR1747200MG

missing translation for 'validMainframeMsg'

4′-Aminoacetophenone, commonly known as Clenbuterol Impurity D, is a related compound of sympathomimetic amine drug clenbuterol. Clenbuterol belongs to a group of compounds referred to as β-2-agonists and is generally used in the treatment of asthma.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
TRUSTED_SUSTAINABILITY

Specifications

CAS 99-92-3
Linear Formula H2NC6H4COCH3
MDL Number MFCD00007896
Melting Point 103°C to 107°C
Boiling Point 293°C
Synonym 4 '-Aminoacetophenone; 4-Acetylaniline
Quantity 200 mg
Formula Weight 135.16
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.