Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Aminoacetophenone 98.0+%, TCI America™
SDP

Catalog No. A025125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A025125G 25 g
A0251100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A025125G Supplier TCI America Supplier No. A025125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 99-92-3
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007896
InChI Key GPRYKVSEZCQIHD-UHFFFAOYSA-N
Synonym 4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino
PubChem CID 7468
IUPAC Name 1-(4-aminophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 107°C
Color Yellow
Boiling Point 293°C
Quantity 25 g
Formula Weight 135.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Aminoacetophenone
RTECSNumber AM5500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.