Learn More
Selleck Chemical LLC 4-Aminoantipyrine S4509-100mg
SDP
Catalog No. 503111155
Encompass_Preferred
4-Aminoantipyrine(Ampyrone) is a metabolite of aminopyrine with analgesic anti-inflammatory and antipyretic properties It is used as a reagent for biochemical reactions producing peroxides or phenols
Catalog No. 50-311-1155 Supplier Selleck Chemical LLC Supplier No. S4509100MG
Add to Cart
Add to Cart
4-Aminoantipyrine(Ampyrone) is a metabolite of aminopyrine with analgesic anti-inflammatory and antipyretic properties It is used as a reagent for biochemical reactions producing peroxides or phenols