Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoantipyrine, ≥98.0%, MilliporeSigma™ Supelco™

Catalog No. 111015640 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5640 100 g
11-101-5639 25 g
11-101-5638 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-5640 Supplier MilliporeSigma Supelco Supplier No. 06800100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00003145
Synonym 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one; Ampyrone

Specifications

Melting Point 105°C to 110°C; 108°C to 110°C
CAS 83-07-8
Linear Formula C11H13N3 O
Quantity 100 g
Solubility Information Water: 0.1 g/mL, clear
Formula Weight 203.24
Percent Purity ≥98.0% (NT); ≥98.0%
RTECSNumber CD2480000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.