Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoantipyrine, 99%, MP Biomedicals™

Catalog No. MP215034310 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215034310 10 g
ICN15034380 100 g
ICN15034383 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215034310 Supplier MP Biomedicals, Inc Supplier No. 0215034310
Add to Cart
Add to Cart

4-Aminoantipyrine, Yellow Powder

  • Reagent for determination of chlorinated phenols, phenylglycosidase, and glucose in the presence of phenol and peroxidase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C11H13N3O
InChI Key RLFWWDJHLFCNIJ-UHFFFAOYSA-N
PubChem CID 2151

Specifications

Color Yellow
Quantity 10 g
Synonym 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone
Molecular Weight (g/mol) 203.25
Physical Form Powder
RTECSNumber CD2480000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.