Learn More
Sigma Aldrich Fine Chemicals Biosciences 4-Aminoantipyrine reagent grade | 83-07-8 | MFCD00003145 | 50G
Catalog No. 501854371
Encompass
4-Aminoantipyrine reagent grade | Purity: >=97% (HPLC) | Mol Wt: 203.24 | 83-07-8 | MFCD00003145 | 50G
Catalog No. 50-185-4371 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A438250G
4-Aminoantipyrine reagent grade | Purity: >=97% (HPLC) | Mol Wt: 203.24 | 83-07-8 | MFCD00003145 | 50G