Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Aminobenzamide | 2835-68-9 | MFCD00007999 | 98.7% | 136.15 g/mol | C7H8N2O | 100 G
SDP

Catalog No. 5000300616
Encompass_Preferred

4-Aminobenzamide (C7H8N2O, CAS 2835-68-9) is a small-molecule biochemical reagent used in life-science research. It serves as an aromatic amide building block in organic synthesis and as a reagent in biochemical assays. The product is supplied as a high-purity solid with recommended storage at 4°C and protection from light; solvent storage conditions are provided for long-term stability.

  • High purity suitable for research applications.
  • White to off-white solid appearance.
  • Molecular weight 136.15 g/mol.
  • Recommended storage at 4°C, protect from light.
  • In solvent: stable at -80°C for months or -20°C for short term.
  • Available in multiple pack sizes including 100 g.

Catalog No. 50-003-00616 Supplier Medchemexpress LLC Supplier No. HYW017041100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Aminobenzamide (C7H8N2O, CAS 2835-68-9) is a small-molecule biochemical reagent used in life-science research. It serves as an aromatic amide building block in organic synthesis and as a reagent in biochemical assays. The product is supplied as a high-purity solid with recommended storage at 4°C and protection from light; solvent storage conditions are provided for long-term stability.

  • High purity suitable for research applications.
  • White to off-white solid appearance.
  • Molecular weight 136.15 g/mol.
  • Recommended storage at 4°C, protect from light.
  • In solvent: stable at -80°C for months or -20°C for short term.
  • Available in multiple pack sizes including 100 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.