Learn More
Apexbio Technology LLC 4-Aminobenzamidine dihydrochloride 2498-50-2 5g
SDP
Catalog No. 5000093912
Encompass_Preferred
4-Aminobenzamidine dihydrochloride (CAS 2498-50-2) is a small molecule inhibitor predominantly targeting serine proteases By binding competitively to the active sites of these enzymes it impedes proteolytic activity thereby modulating protease-mediated biological processes This compound is widely utilized in biochemical and cell-based assays to investigate the roles of serine proteases in various physiological and pathological contexts Its application extends to research on diseases characterized by dysregulated protease activity including certain cancers and inflammatory disorders where it serves as a tool for drug development and mechanistic studies
Catalog No. 50-000-93912 Supplier Apexbio Technology LLC Supplier No. A99115000
4-Aminobenzamidine dihydrochloride (CAS 2498-50-2) is a small molecule inhibitor predominantly targeting serine proteases By binding competitively to the active sites of these enzymes it impedes proteolytic activity thereby modulating protease-mediated biological processes This compound is widely utilized in biochemical and cell-based assays to investigate the roles of serine proteases in various physiological and pathological contexts Its application extends to research on diseases characterized by dysregulated protease activity including certain cancers and inflammatory disorders where it serves as a tool for drug development and mechanistic studies