Learn More
Medchemexpress LLC 4-aminobenzamidine (dihydrochloride) | 2498-50-2 | MFCD00013001 | 99.9% | 208.09 g/mol | C7H11Cl2N3 | 5 G
SDP
Catalog No. 5000302029
Encompass_Preferred
4-Aminobenzamidine dihydrochloride is a small-molecule serine protease inhibitor used in biochemical and pharmacological research. It functions as a strong trypsin inhibitor and a urokinase-type plasminogen activator (uPA) inhibitor, and is commonly employed in studies of cardiovascular function and tumor biology. The compound is supplied as the dihydrochloride salt in solid and solution formats for laboratory use.
  • Strong trypsin inhibitor and uPA inhibitor (Ki ≈ 82 μM).
  • High purity (~99.9%) supported by certificate of analysis.
  • Molecular weight 208.09 g/mol and formula C7H11Cl2N3.
  • Available as solid (including 5 g) and as 10 mM solutions in DMSO.
  • Provided with COA and SDS for quality and safety information.
Catalog No. 50-003-02029 Supplier Medchemexpress LLC Supplier No. HYW0187235G
Add to Cart
Add to Cart
4-Aminobenzamidine dihydrochloride is a small-molecule serine protease inhibitor used in biochemical and pharmacological research. It functions as a strong trypsin inhibitor and a urokinase-type plasminogen activator (uPA) inhibitor, and is commonly employed in studies of cardiovascular function and tumor biology. The compound is supplied as the dihydrochloride salt in solid and solution formats for laboratory use.
  • Strong trypsin inhibitor and uPA inhibitor (Ki ≈ 82 μM).
  • High purity (~99.9%) supported by certificate of analysis.
  • Molecular weight 208.09 g/mol and formula C7H11Cl2N3.
  • Available as solid (including 5 g) and as 10 mM solutions in DMSO.
  • Provided with COA and SDS for quality and safety information.