Learn More
Medchemexpress LLC 4-Aminobenzo-12-crown-4 | 78554-68-4 | 98.0% | C12H17NO4 | 1 G
SDP
Catalog No. 5000433321
Encompass_Preferred
4-Aminobenzo-12-crown-4 is a solid chemical compound primarily utilized as a drug intermediate for the synthesis of various active pharmaceutical ingredients. It is stable under recommended storage conditions and finds application in laboratory research and the broader manufacturing of chemical substances.
  • Drug intermediate for synthesis of active compounds
  • Suitable for laboratory research
  • Used in manufacturing of substances
  • Stable under recommended storage conditions
  • Solid physical appearance
  • Recommended storage at 4°C, protected from light
Catalog No. 50-004-33321 Supplier Medchemexpress LLC Supplier No. HY783341G
4-Aminobenzo-12-crown-4 is a solid chemical compound primarily utilized as a drug intermediate for the synthesis of various active pharmaceutical ingredients. It is stable under recommended storage conditions and finds application in laboratory research and the broader manufacturing of chemical substances.
  • Drug intermediate for synthesis of active compounds
  • Suitable for laboratory research
  • Used in manufacturing of substances
  • Stable under recommended storage conditions
  • Solid physical appearance
  • Recommended storage at 4°C, protected from light