Learn More
Medchemexpress LLC 4-Aminobenzoic acid | 150-13-0 | 99.93% | 10 G
SDP
Catalog No. 5000407510
Encompass_Preferred
4-Aminobenzoic acid is an intermediate in the synthesis of folic acid by bacteria, plants, and fungi. It possesses antibacterial and plant growth regulating properties. It has also been used as an active constituent in sunscreens to protect against UVB irradiation, acting as a scavenger of reactive oxygen species. Derivatives of 4-Aminobenzoic acid have been associated with cognition in Charles foster rats.
  • Acts as a microbial and human endogenous metabolite.
  • Shows antibacterial activity against *Listeria monocytogenes*, *Salmonella enteritidis*, and *E. coli*.
  • Promotes resistance against plant bacterial and viral pathogens.
  • Functions as a plant growth modulator.
  • Can act as a scavenger of reactive oxygen species.
  • Associated with cognition in Charles foster rats.
Catalog No. 50-004-07510 Supplier Medchemexpress LLC Supplier No. HYB100810G
4-Aminobenzoic acid is an intermediate in the synthesis of folic acid by bacteria, plants, and fungi. It possesses antibacterial and plant growth regulating properties. It has also been used as an active constituent in sunscreens to protect against UVB irradiation, acting as a scavenger of reactive oxygen species. Derivatives of 4-Aminobenzoic acid have been associated with cognition in Charles foster rats.
  • Acts as a microbial and human endogenous metabolite.
  • Shows antibacterial activity against *Listeria monocytogenes*, *Salmonella enteritidis*, and *E. coli*.
  • Promotes resistance against plant bacterial and viral pathogens.
  • Functions as a plant growth modulator.
  • Can act as a scavenger of reactive oxygen species.
  • Associated with cognition in Charles foster rats.