Learn More
Medchemexpress LLC 4-Aminobenzoic acid | 150-13-0 | 99.93% | 100 G
SDP
Catalog No. 5000407487
Encompass_Preferred
4-Aminobenzoic acid, also known as PABA, is an intermediate involved in the synthesis of folic acid in bacteria, plants, and fungi. It possesses antibacterial and plant growth-regulating properties. With a molecular weight of 137.14 and a chemical formula of C7H7NO2, this compound is supplied as an off-white to light brown solid.
  • Intermediate in folic acid synthesis
  • Exhibits antibacterial and plant growth-regulating properties
  • Direct antibacterial activity against *Listeria monocytogenes*, *Salmonella enteritidis*, and *E. coli*
  • Utilized in sunscreens to protect against UVB irradiation
  • Promotes resistance against plant bacterial and viral pathogens
  • Acts as a plant growth modulator
  • Derivatives associated with cognition in Charles Foster rats
Catalog No. 50-004-07487 Supplier Medchemexpress LLC Supplier No. HYB1008100G
4-Aminobenzoic acid, also known as PABA, is an intermediate involved in the synthesis of folic acid in bacteria, plants, and fungi. It possesses antibacterial and plant growth-regulating properties. With a molecular weight of 137.14 and a chemical formula of C7H7NO2, this compound is supplied as an off-white to light brown solid.
  • Intermediate in folic acid synthesis
  • Exhibits antibacterial and plant growth-regulating properties
  • Direct antibacterial activity against *Listeria monocytogenes*, *Salmonella enteritidis*, and *E. coli*
  • Utilized in sunscreens to protect against UVB irradiation
  • Promotes resistance against plant bacterial and viral pathogens
  • Acts as a plant growth modulator
  • Derivatives associated with cognition in Charles Foster rats