Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Aminobenzoic Acid

Catalog No. A9878100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A9878100G 25 g
A987825G 5 g
A98785G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A9878100G Supplier Sigma Aldrich Supplier No. A9878100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Aminobenzoic Acid

Specifications

CAS 150-13-0
Melting Point 187°C to 189°C
Density 1.374 g/mL at 25°C
Molecular Formula C7H7NO2
Linear Formula H2NC6H4CO2H
Quantity 25 g
MDL Number MFCD00007894
Synonym para-Aminobenzoic acid; PABA; Vitamin Bx; Vitamin H1
Molecular Weight (g/mol) 137.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.