Learn More
Medchemexpress LLC 4-Aminobenzoic acid-d4 | 350820-01-8 | 99.4% | 5 MG
SDP
Catalog No. 5000407476
Encompass_Preferred
4-Aminobenzoic acid-d4 is the deuterium labeled 4-Aminobenzoic acid, an intermediate in the synthesis of folate by bacteria, plants, and fungi. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07476 Supplier Medchemexpress LLC Supplier No. HYB1008S5MG
4-Aminobenzoic acid-d4 is the deuterium labeled 4-Aminobenzoic acid, an intermediate in the synthesis of folate by bacteria, plants, and fungi. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS