Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Aminobenzoic acid potassium | 138-84-1 | MFCD00064911 | 99.96% | 5 G
SDP

Catalog No. 5000407478
Encompass_Preferred

4-Aminobenzoic acid potassium, also known as PABA potassium, is recognized as a modifier of catecholamine oxidation reactions. It shows promise for research related to Parkinson's disease and hypertension, functioning as both a microbial and human endogenous metabolite.

  • Modifier of catecholamine oxidation reactions
  • Promising for Parkinson's disease and hypertension research
  • Target: microbial and human endogenous metabolite
  • Appearance: solid
  • Color: white to off-white
  • Solubility: 100 mg/mL in H2O

Catalog No. 50-004-07478 Supplier Medchemexpress LLC Supplier No. HYB1008A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Aminobenzoic acid potassium, also known as PABA potassium, is recognized as a modifier of catecholamine oxidation reactions. It shows promise for research related to Parkinson's disease and hypertension, functioning as both a microbial and human endogenous metabolite.

  • Modifier of catecholamine oxidation reactions
  • Promising for Parkinson's disease and hypertension research
  • Target: microbial and human endogenous metabolite
  • Appearance: solid
  • Color: white to off-white
  • Solubility: 100 mg/mL in H2O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.