Learn More
Medchemexpress LLC 4-Aminobenzonitrile-d4 | 1331866-32-0 | 98% | 122.16 | 1 MG
SDP
Catalog No. 5000404885
Encompass_Preferred
4-Aminobenzonitrile-d4 is the deuterium labeled 4-Aminobenzonitrile. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, achieved by incorporating stable heavy isotopes of hydrogen into drug molecules, has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-04885 Supplier Medchemexpress LLC Supplier No. HY41680S1MG
4-Aminobenzonitrile-d4 is the deuterium labeled 4-Aminobenzonitrile. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, achieved by incorporating stable heavy isotopes of hydrogen into drug molecules, has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS