Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-aminobenzophenone | 1137-41-3 | MFCD00007895 | 99.9% | 197.23 g/mol | C13H11NO | 25 G
SDP

Catalog No. 5000208837
Encompass_Preferred

4-Aminobenzophenone is an aromatic benzophenone derivative that functions as a photosensitizer and an intermediate in organic synthesis. It is supplied as a light yellow to brown solid with high purity and is commonly used in photochemical applications and research.

  • High purity (99.91%).
  • Light yellow to brown solid appearance.
  • Molecular formula C13H11NO, molecular weight 197.23 g/mol.
  • Used as a photosensitizer and synthetic intermediate.
  • Available in small laboratory pack sizes, such as 25 g and 50 g.

Catalog No. 50-002-08837 Supplier Medchemexpress LLC Supplier No. 25GHYW076903
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Aminobenzophenone is an aromatic benzophenone derivative that functions as a photosensitizer and an intermediate in organic synthesis. It is supplied as a light yellow to brown solid with high purity and is commonly used in photochemical applications and research.

  • High purity (99.91%).
  • Light yellow to brown solid appearance.
  • Molecular formula C13H11NO, molecular weight 197.23 g/mol.
  • Used as a photosensitizer and synthetic intermediate.
  • Available in small laboratory pack sizes, such as 25 g and 50 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.