Learn More
Medchemexpress LLC 4-Aminobenzothioamide | 4714-67-4 | MFCD00040927 | 98.7% | C7H8N2S | 1 ML
SDP
Catalog No. 5000434171
Encompass_Preferred
4-Aminobenzothioamide is a small H2S donor. It has potential for the research of myocardial ischemia-reperfusion injury.
  • Small H2S donor
  • Potential for research of myocardial ischemia-reperfusion injury
Catalog No. 50-004-34171 Supplier Medchemexpress LLC Supplier No. HY7614410MM/1ML
Add to Cart
Add to Cart
4-Aminobenzothioamide is a small H2S donor. It has potential for the research of myocardial ischemia-reperfusion injury.
  • Small H2S donor
  • Potential for research of myocardial ischemia-reperfusion injury