Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzyl alcohol, 98%

Catalog No. AAA1413006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1413006 5 g
AAA1413014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1413006 Supplier Thermo Scientific Chemicals Supplier No. A1413006
  • $131.00
    $128.65 / Each of 1

    Save $2.35

    2% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 623-04-1 | C7H9NO | 123.16 g/mol

4-Aminobenzyl alcohol is used in the synthesis of 4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino}benzyl ester. It is also used as an intermediate in organic synthesis and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzyl alcohol is used in the synthesis of 4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino}benzyl ester. It is also used as an intermediate in organic synthesis and pharmaceutical products.

Solubility
Soluble in alcohol, ether and benzene. Partially soluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents and strong acids. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-04-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00014782
InChI Key AXKGIPZJYUNAIW-UHFFFAOYSA-N
Synonym 4-aminobenzyl alcohol, 4-aminophenyl methanol, 4-aminobenzylalcohol, 4-hydroxymethyl aniline, p-aminobenzyl alcohol, p-aminobenzylalcohol, benzenemethanol, 4-amino, benzyl alcohol, p-amino, 4-amino-benzylalcohol, pubchem7480
PubChem CID 69331
IUPAC Name (4-aminophenyl)methanol
SMILES NC1=CC=C(CO)C=C1

Specifications

Melting Point 59°C to 63°C
Quantity 5 g
Beilstein 2078680
Sensitivity Air and light sensitive
Solubility Information Soluble in alcohol,ether and benzene. Partially soluble in water.
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 4-Aminobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-767-5
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.