Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Aminobenzyl)phosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. A30721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30721G Supplier TCI America Supplier No. A30721G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5424-27-1
Molecular Formula C7H10NO3P
Molecular Weight (g/mol) 187.135
MDL Number MFCD00070204
InChI Key NEKHKXMBGWNTOO-UHFFFAOYSA-N
PubChem CID 223978
IUPAC Name (4-aminophenyl)methylphosphonic acid
SMILES C1=CC(=CC=C1CP(=O)(O)O)N

Specifications

Melting Point 325°C
Color White-Yellow
Quantity 1 g
Formula Weight 187.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Aminobenzyl)phosphonic Acid
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.