Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobiphenyl, 98%

Catalog No. p-7036938
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5542403 1 g
AAH5542406 5 g
AAH5542414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5542403 Supplier Thermo Scientific Chemicals Supplier No. H5542403
Only null left
Add to Cart

CAS: 92-67-1 | C12H11N | 169.227 g/mol

It is used to manufacture azo dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to manufacture azo dyes.

Solubility
Soluble in Dichloromethane, DMSO and Methanol

Notes
Store at -20°C. Avoid heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-67-1
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
MDL Number MFCD00007879
InChI Key DMVOXQPQNTYEKQ-UHFFFAOYSA-N
Synonym 4-aminobiphenyl, 4-aminodiphenyl, 4-biphenylamine, biphenyl-4-ylamine, 1,1'-biphenyl-4-amine, biphenyl-4-amine, xenylamine, 4-biphenylylamine, p-phenylaniline, p-aminobiphenyl
PubChem CID 7102
ChEBI CHEBI:1784
IUPAC Name 4-phenylaniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N

Specifications

Melting Point 52°C to 54°C
Boiling Point 191°C (15 mmHg)
Quantity 1 g
UN Number UN3077
Beilstein 386533
Merck Index 14,1235
Solubility Information Soluble in Dichloromethane,DMSO and Methanol
Formula Weight 169.23
Percent Purity 98%
Chemical Name or Material 4-Aminobiphenyl
Hazard Category H302-H350
Hazard Statement GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-177-1
RTECSNumber DU8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.