Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobiphenyl, MilliporeSigma™ Supelco™

Catalog No. 111019840 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9840 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9840 Supplier MilliporeSigma Supelco Supplier No. 31598250MG
Add to Cart
Add to Cart

4-Aminobiphenyl is classified under the aromatic amine group of compounds.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 842°F
CAS 92-67-1
Flash Point 113°C - Closed Cup
Physical Form Neat
Linear Formula C6H5C6H4 NH2
MDL Number MFCD00007879
Melting Point 52°C to 54°C
Boiling Point 191°C (15 mmHg)
Synonym 4-Aminodiphenyl; 4-Biphenylamine; 4-Biphenylylamine; 4-Phenylaniline; Biphenyl-4-ylamine; NSC 7660; Xenylamine
Type Analytical Standard
Quantity 250 mg
Formula Weight 169.22
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.