Learn More
Medchemexpress LLC 4-Aminobutan-1-ol | 13325-10-5 | 89.14 | 10 G
SDP
Catalog No. 5000447826
Encompass_Preferred
4-Aminobutan-1-ol is a chemical compound characterized by the presence of both an amino group and a hydroxyl group. This versatile compound is primarily utilized as a key intermediate in the synthesis of various other active compounds.
  • Appears as a solid below 16°C and a liquid above 18°C
  • Colorless to off-white appearance
  • Stable under recommended storage conditions
  • Suitable for research use only
Catalog No. 50-004-47826 Supplier Medchemexpress LLC Supplier No. HYW00204110G
4-Aminobutan-1-ol is a chemical compound characterized by the presence of both an amino group and a hydroxyl group. This versatile compound is primarily utilized as a key intermediate in the synthesis of various other active compounds.
  • Appears as a solid below 16°C and a liquid above 18°C
  • Colorless to off-white appearance
  • Stable under recommended storage conditions
  • Suitable for research use only