Learn More
Medchemexpress LLC 4-Aminobutan-1-ol | 13325-10-5 | MFCD00008230 | 89.14 | 10MM 1ML

Supplier: Medchemexpress LLC HYW00204110MM/1ML
4-Aminobutan-1-ol is a compound featuring both an amino group and a hydroxyl group. It is an important intermediate that can be used for the synthesis of other active compounds.
- Contains both amino and hydroxyl groups
- Important intermediate for synthesis
- Used for synthesizing other active compounds
- Appearance: colorless to off-white solid below 16°C, liquid above 18°C
- Storage: 4°C, protected from light
- Storage in solvent: -80°C for 6 months; -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.