Learn More
AdipoGen 4-Aminobutanoic acid
SDP
Supplier:  AdipoGen CDXG0043G500
Encompass_Preferred
Chemical. CAS 56-12-2. Formula C4H9NO2. MW 103.12. Synthetic. An amino acid that functions as the major inhibitory neurotransmitter in the mammalian central nervous system and also functions as a neuromodulator in some peripheral tissues. GABAA and GABAB receptor agonist that increases Cl− conductance. GABA acts at inhibitory synapses in the brain by binding to specific transmembrane receptors in the plasma membrane of both pre- and postsynaptic neuronal processes. This binding causes the opening of ion channels to allow the flow of either negatively charged chloride ions into the cell or positively charged potassium ions out of the cell. This action results in a negative change in the transmembrane potential, usually causing hyperpolarization. GABAergic neurons are involved in myorelaxation, anxiolytic treatment, sedation and anaesthetics. GABA can also influence heart rate and blood pressure and function as a immunomodulator.
Catalog No. 50-256-0828