Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Aminobutylphosphonic acid | 35622-27-6 | MFCD00063426 | 98.0% | 153.12 g/mol | C4H12NO3P | 250 MG
SDP

Catalog No. 5000302737
Encompass_Preferred

4-Aminobutylphosphonic acid is a phosphonic acid-containing amino acid derivative intended for research use where stable phosphonate analogs of amino acids are required. Supplied as a solid powder, it is suitable for biochemical and enzymatic studies involving phosphonate chemistry.

  • High purity: 98.0%.
  • Molecular formula C4H12NO3P; molecular weight 153.12 g/mol.
  • Solid powder appearance, typically white to gray.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available pack sizes include 250 mg, 1 g, 5 g, 10 g, and 50 g.
  • Useful as a phosphonate analog in biochemical and enzymatic research.

Catalog No. 50-003-02737 Supplier Medchemexpress LLC Supplier No. 250MGHYW018502
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Aminobutylphosphonic acid is a phosphonic acid-containing amino acid derivative intended for research use where stable phosphonate analogs of amino acids are required. Supplied as a solid powder, it is suitable for biochemical and enzymatic studies involving phosphonate chemistry.

  • High purity: 98.0%.
  • Molecular formula C4H12NO3P; molecular weight 153.12 g/mol.
  • Solid powder appearance, typically white to gray.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available pack sizes include 250 mg, 1 g, 5 g, 10 g, and 50 g.
  • Useful as a phosphonate analog in biochemical and enzymatic research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.