Learn More
Sigma Aldrich 4-Aminobutyraldehyde diethyl acetal
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Traditionally challenging to access, this hindered amine provides a chemically differentiated building block for organic synthesis and medicinal chemistry, specifically for preparation of drug candidates containing hindered amine motifs
- Additionally, this and over 80 other sterically hindered secondary amines in the sigma-aldrich portfolio provide structurally distinct screening compounds
- Screen several through aldrich market select
- This hindered amine was accessed via a recent publication reported by baran and coworkers using an innovative hydroamination method.product of kemxtree
Specifications
Specifications
| Density | 0.933 g/mL (at 25°C (literature)) |
| Boiling Point | 196°C (lit.) |
| Molecular Formula | C8H19NO2 |
| Refractive Index | n20/D 1.428 (literature) |
| Linear Formula | H2N(CH2)3 CH(OC2H5)2 |
| MDL Number | MFCD00008227 |
| Quantity | 250 mg |
| Synonym | 4,4-Diethoxybutylamine |
| Molecular Weight (g/mol) | 161.24 |
| Percent Purity | 90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.