Learn More
Medchemexpress LLC 4-(aminocarbonyl)-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-phenylalanine | 204716-17-6 | 99.3% | 430.45 | C25H22N2O5 | 100 MG
SDP
Catalog No. 5000210853
Encompass_Preferred
Fmoc-Phe(4-CONH2)-OH is an Fmoc-protected derivative of para-carbamoyl phenylalanine used as an amino acid building block in peptide synthesis and related research applications. It is supplied in multiple pre-weighed sizes and is documented with a datasheet, certificate of analysis, and safety data sheet.
  • Fmoc-protected amino acid suitable for solid-phase peptide synthesis.
  • High purity for research use.
  • Available in multiple pre-weighed sizes for flexibility.
  • Documented with datasheet, COA, and SDS for quality and safety information.
  • Stable as a solid at room temperature for extended storage.
Catalog No. 50-002-10853 Supplier Medchemexpress LLC Supplier No. HYW048673100MG
Fmoc-Phe(4-CONH2)-OH is an Fmoc-protected derivative of para-carbamoyl phenylalanine used as an amino acid building block in peptide synthesis and related research applications. It is supplied in multiple pre-weighed sizes and is documented with a datasheet, certificate of analysis, and safety data sheet.
  • Fmoc-protected amino acid suitable for solid-phase peptide synthesis.
  • High purity for research use.
  • Available in multiple pre-weighed sizes for flexibility.
  • Documented with datasheet, COA, and SDS for quality and safety information.
  • Stable as a solid at room temperature for extended storage.