Learn More
Description
- N,n-dimethylformamide dineopentyl acetal was used in the esterification of nα-9-fluorenylmethyloxycarbonylamino acids(fmoc)
- It was used in the synthesis of 1,3-dialkyl, benzyl and cyclohexyl barbiturate derivatives
- It was used as reagent during the synthesis of L-serine and L-cystine stereospecifically labeled with deuterium at the α-position
Specifications
Specifications
| Molecular Formula | C6H11NO |
| Linear Formula | C6H11NO |
| MDL Number | MFCD06659766 |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 113.16 |
| Biological Activity | Respiratory System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.