Learn More
Selleck Chemical LLC 4-Aminohippuric Acid S2883-200mg
SDP
Catalog No. 503110091
Encompass_Preferred
4-Aminohippuric acid is a typical substrate of organic anion transport systems
Catalog No. 50-311-0091 Supplier Selleck Chemical LLC Supplier No. S2883200MG
4-Aminohippuric acid is a typical substrate of organic anion transport systems