Learn More
Cayman Chemical 4-Aminohippuric Acid, Purity: ≥98%, CAS: 61-78-9, 10 g
SDP
Catalog No. 502994755
Encompass_Preferred
A substrate for OAT1 (Km = 14.3 µM), NPT1 (Km = 2.66 mM), and MDR2 (Km = 880 µM); a glycine amide form of 4-aminobenzoic acid
Catalog No. 50-299-4755 Supplier Cayman Chemical Supplier No. 2372610 G
A substrate for OAT1 (Km = 14.3 µM), NPT1 (Km = 2.66 mM), and MDR2 (Km = 880 µM); a glycine amide form of 4-aminobenzoic acid