Learn More
Apexbio Technology LLC 4-Aminohippuric Acid 61-78-9 200mg
SDP
Catalog No. 502240757
Encompass_Preferred
4-Aminohippuric acid (CAS 61-78-9) is a small organic molecule recognized as a prototypical substrate for organic anion transporters (OATs) It interacts selectively with renal tubular transport systems facilitating studies on tubular secretion and renal excretory function In biomedical research 4-aminohippuric acid is commonly utilized to assess the activity and specificity of organic anion transport mechanisms particularly in kidney physiology and pharmacokinetics Its application enables quantitative evaluation of renal plasma flow and investigation of transporter-mediated drug interactions
This item is not returnable. View return policy
Catalog No. 50-224-0757 Supplier Apexbio Technology LLC Supplier No. B1873200
4-Aminohippuric acid (CAS 61-78-9) is a small organic molecule recognized as a prototypical substrate for organic anion transporters (OATs) It interacts selectively with renal tubular transport systems facilitating studies on tubular secretion and renal excretory function In biomedical research 4-aminohippuric acid is commonly utilized to assess the activity and specificity of organic anion transport mechanisms particularly in kidney physiology and pharmacokinetics Its application enables quantitative evaluation of renal plasma flow and investigation of transporter-mediated drug interactions