Learn More
Medchemexpress LLC 4-aminohippuric acid | 61-78-9 | MFCD00007890 | 100.0% | 194.19 g/mol | C9H10N2O3 | 10 MG
SDP
Catalog No. 5000297173
Encompass_Preferred
4-Aminohippuric acid (p-aminohippuric acid) analytical standard supplied in a small 10 mg package for research and analytical applications. It is used as a coordination ligand for metal ions, and as an established substrate for studies of organic anion transport and renal plasma flow measurement.
  • Analytical standard suitable for chromatography and mass spectrometry.
  • High purity for analytical use.
  • Small-scale packaging convenient for method development and standards preparation.
  • Useful as a coordination ligand and research reagent in materials and biochemical studies.
Catalog No. 50-002-97173 Supplier Medchemexpress LLC Supplier No. HYB1306R10MG
4-Aminohippuric acid (p-aminohippuric acid) analytical standard supplied in a small 10 mg package for research and analytical applications. It is used as a coordination ligand for metal ions, and as an established substrate for studies of organic anion transport and renal plasma flow measurement.
  • Analytical standard suitable for chromatography and mass spectrometry.
  • High purity for analytical use.
  • Small-scale packaging convenient for method development and standards preparation.
  • Useful as a coordination ligand and research reagent in materials and biochemical studies.