Learn More
Medchemexpress LLC 4-Aminohippuric acid | 61-78-9 | MFCD00007890 | 99.9% | 100 MG
SDP
Catalog No. 5000411544
Encompass_Preferred
4-Aminohippuric acid (p-Aminohippuric acid) (Standard) is an analytical standard for research and analytical applications. It serves as a coordination ligand for metal ions (e.g., Cu2+, Fe3+, Hg2+) and a functionalization reagent for nanomaterials.
  • Coordinates with metal ions or modifies surfaces like carbon nanotubes and gold nanoparticles via amino and carboxyl groups.
  • Forms stable complexes with metal ions or aids nanomaterial synthesis as a reducing agent/stabilizer.
  • Used to construct sensitive electrochemical or colorimetric sensors for heavy metal ions in environmental and biological samples.
  • May serve as a biomarker for attention-deficit/hyperactivity disorder (ADHD).
  • An analytical standard for assays, commonly used in HPLC, GC, and MS research experiments.
Catalog No. 50-004-11544 Supplier Medchemexpress LLC Supplier No. HYB1306R100MG
4-Aminohippuric acid (p-Aminohippuric acid) (Standard) is an analytical standard for research and analytical applications. It serves as a coordination ligand for metal ions (e.g., Cu2+, Fe3+, Hg2+) and a functionalization reagent for nanomaterials.
  • Coordinates with metal ions or modifies surfaces like carbon nanotubes and gold nanoparticles via amino and carboxyl groups.
  • Forms stable complexes with metal ions or aids nanomaterial synthesis as a reducing agent/stabilizer.
  • Used to construct sensitive electrochemical or colorimetric sensors for heavy metal ions in environmental and biological samples.
  • May serve as a biomarker for attention-deficit/hyperactivity disorder (ADHD).
  • An analytical standard for assays, commonly used in HPLC, GC, and MS research experiments.